missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenethyl Alcohol, TRC

Product Code. 30310230
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30310230 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310230 Supplier Toronto Research Chemicals Supplier No. TRCA58504510G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Aminophenethyl Alcohol
CAS 104-10-9
Molecular Formula C8H11NO
Quantity 10 g
Synonym 4-Aminobenzeneethanol, 2-(4-Aminophenyl)-1-ethanol, 2-(4-Aminophenyl)ethanol, 2-(4-Aminophenyl)ethyl alcohol, 2-(p-Aminophenyl)ethanol
InChI Formula InChI=1S/C8H11NO/c9-8-3-1-7(2-4-8)5-6-10/h1-4,10H,5-6,9H2
SMILES Nc1ccc(CCO)cc1
IUPAC Name 2-(4-aminophenyl)ethanol
Molecular Weight (g/mol) 137.18
Formula Weight 137.0841
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.