missing translation for 'onlineSavingsMsg'
Learn More

4-Aminonicotinic acid dihydrochloride, Thermo Scientific™

Product Code. 10341660
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10341660 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10341660 Supplier Thermo Scientific Maybridge Supplier No. MAY00123DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 1138011-19-4
Packaging Amber Glass Bottle
Molecular Formula C6H8Cl2N2O2
Quantity 5 g
Synonym 4-aminonicotinic acid dihydrochloride, 4-aminopyridine-3-carboxylic acid dihydrochloride, c6h6n2o2.2hcl, 4-aminopyridine-3-carboxylic acid, chloride, chloride, 4-aminopyridine-3-carboxylic acid-hydrogen chloride 1/2
InChI Key ZCKYJHVCJAUVAE-UHFFFAOYSA-N
SMILES C1=CN=CC(=C1N)C(=O)O.Cl.Cl
IUPAC Name 4-aminopyridine-3-carboxylic acid;dihydrochloride
Molecular Weight (g/mol) 211.042
PubChem CID 45594305
Formula Weight 211.05
Percent Purity 97%
Chemical Name or Material 4-Aminonicotinic acid dihydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.