missing translation for 'onlineSavingsMsg'
Learn More

4-Aminonaphthalene-1-sulfonic acid, 98%, Thermo Scientific™

Catalog Number 11464867
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
84-86-6
C10H9NO3S
223.25
MFCD00004027
NRZRRZAVMCAKEP-UHFFFAOYSA-N
naphthionic acid, 4-amino-1-naphthalenesulfonic acid, piria's acid, 1-naphthylamine-4-sulfonic acid, naphthionsaeure, 1,4-naphthionic acid, sulfonaphtin, 1-amino-4-sulfonaphthalene, usaf m-5, 1-aminonaphthalene-4-sulfonic acid
6790
CHEBI:38219
4-aminonaphthalene-1-sulfonic acid
NC1=C2C=CC=CC2=C(C=C1)S(O)(=O)=O
This item is not returnable. View return policy
84-86-6
C10H9NO3S
223.25
MFCD00004027
NRZRRZAVMCAKEP-UHFFFAOYSA-N
naphthionic acid, 4-amino-1-naphthalenesulfonic acid, piria's acid, 1-naphthylamine-4-sulfonic acid, naphthionsaeure, 1,4-naphthionic acid, sulfonaphtin, 1-amino-4-sulfonaphthalene, usaf m-5, 1-aminonaphthalene-4-sulfonic acid
6790
CHEBI:38219
4-aminonaphthalene-1-sulfonic acid
NC1=C2C=CC=CC2=C(C=C1)S(O)(=O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Amino-1-naphthalenesulfonic acid has potent therapeutic applications. Its sodium salt has been used as non-toxic hemostatic. It is also a metabolite of azo dyes.

Solubility
Sparingly soluble in dichloromethane. Soluble in aqueous base.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point >300°C
Quantity 25 g
UN Number UN2585
Beilstein 1971299
Merck Index 14,6403
Solubility Information Sparingly soluble in dichloromethane. Soluble in aqueous base.
Formula Weight 223.25
Percent Purity 98%
Chemical Name or Material 4-Aminonaphthalene-1-sulfonic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'