missing translation for 'onlineSavingsMsg'
Learn More

4-Aminomethyltetrahydropyran, 97%

Product Code. 10126703 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Unit Size:
1g
250mg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10126703 5 g 5g
10184693 250 mg 250mg
10143813 1 g 1g
11976201 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10126703 Supplier Thermo Scientific Acros Supplier No. 385210050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 130290-79-8 | C6H13NO | 115.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130290-79-8
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.17
MDL Number MFCD02179435
InChI Key IPBPLHNLRKRLPJ-UHFFFAOYSA-N
Synonym 4-aminomethyltetrahydropyran, 4-aminomethyl tetrahydro-2h-pyran, tetrahydro-2h-pyran-4-yl methanamine, 4-aminomethyl tetrahydropyran, 1-tetrahydro-2h-pyran-4-ylmethanamine, tetrahydropyran-4-yl methylamine, c-tetrahydro-pyran-4-yl-methylamine, tetrahydro-2h-pyran-4-yl methylamine, 2h-pyran-4-methanamine, tetrahydro, oxan-4-yl methanamine
PubChem CID 2773210
IUPAC Name oxan-4-ylmethanamine
SMILES C1COCCC1CN

Specifications

Density 1.0200g/mL
Boiling Point 75.0°C to 76.0°C (18.0 mmHg)
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4640 to 1.4660
Quantity 5 g
Specific Gravity 1.02
Formula Weight 115.17
Percent Purity 97%
Chemical Name or Material 4-Aminomethyltetrahydropyran
compliance-icons
Product Identifier
  • 1-tetrahydro-2H-pyran-4-ylmethanamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.