missing translation for 'onlineSavingsMsg'
Learn More

4-Aminomethylbenzoic Acid, Mikromol™

Product Code. 17962216
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17962216 100 mg 100mg
17943136 25 mg 25mg
30463862 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17962216 Supplier Mikromol Supplier No. MM0330.04
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Aminomethylbenzoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Aminomethylbenzoic Acid
CAS 56-91-7
Analyte or Component Names 4-Aminomethylbenzoic Acid
Shipping Condition Room Temperature
Molecular Formula C8 H9 N O2
Certifications/Compliance ISO 17025
Synonym Tranexamic Acid Imp. D (EP), 4-Aminomethylbenzoic Acid, Benzoic acid, 4-(aminomethyl)-, p-Toluic acid, alpha-amino- (6CI,8CI), 4-(Aminomethyl)benzoic acid, 4-Carboxybenzylamine, Benzylamine-4-carboxylic acid, Gumbix, NSC 41629, PAMBA, Styptopur, p-(Aminomethyl)benzoic acid, p-Carboxybenzylamine, alpha-Amino-p-toluic acid, Tranexamic Acid Impurity D
InChI Formula InChI=1S/C8H9NO2/c9-5-6-1-3-7(4-2-6)8(10)11/h1-4H,5,9H2,(H,10,11)
SMILES NCc1ccc(cc1)C(=O)O
IUPAC Name 4-(aminomethyl)benzoic acid
Molecular Weight (g/mol) 151.16
Quantity 100 mg
Formula Weight 151.0633 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.