missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)pyridine, 98%

Product Code. p-4813003 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10275004 25 g 25g
10326304 100 g 100g
10235054 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10275004 Supplier Thermo Scientific Acros Supplier No. 382080250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3731-53-1 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3731-53-1
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
InChI Key TXQWFIVRZNOPCK-UHFFFAOYSA-N
Synonym 4-pyridinemethaneamine, 4-aminomethyl pyridine, 4-pyridinemethanamine, 4-pyridylmethylamine, 4-picolylamine, 4-aminomethylpyridine, 4-picolinylamine, 4-pyridinemethylamine, 1-pyridin-4-ylmethanamine, 4-picolinamine
PubChem CID 77317
IUPAC Name pyridin-4-ylmethanamine
SMILES C1=CN=CC=C1CN

Specifications

Melting Point -8.0°C
Density 1.0650g/mL
Boiling Point 228.0°C to 230.0°C
Flash Point 108°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5515
Quantity 25 g
Specific Gravity 1.065
Formula Weight 108.14
Percent Purity 98%
Chemical Name or Material 4-(Aminomethyl)pyridine
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Aminomethyl)pyridine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.