missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)pyridine, 97+%

Product Code. 11392476
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392476 100 g 100g
11382476 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392476 Supplier Thermo Scientific Alfa Aesar Supplier No. L06345.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3731-53-1 | C6H8N2 | 108.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3731-53-1
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006449
InChI Key TXQWFIVRZNOPCK-UHFFFAOYSA-N
Synonym 4-pyridinemethaneamine, 4-aminomethyl pyridine, 4-pyridinemethanamine, 4-pyridylmethylamine, 4-picolylamine, 4-aminomethylpyridine, 4-picolinylamine, 4-pyridinemethylamine, 1-pyridin-4-ylmethanamine, 4-picolinamine
PubChem CID 77317
IUPAC Name pyridin-4-ylmethanamine
SMILES C1=CN=CC=C1CN

Specifications

Melting Point -8°C
Density 1.069
Boiling Point 230°C
Flash Point 108°C (226°F)
Odor Amine-like
Refractive Index 1.5515
Quantity 100 g
UN Number UN2735
Beilstein 108057
Sensitivity Air Sensitive
Formula Weight 108.14
Percent Purity ≥97%
Chemical Name or Material 4-(Aminomethyl)pyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.