missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)-N,N-dimethyltetrahydro-2H-pyran-4-amine, 95%, Thermo Scientific™

Product Code. 10554984
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10554984 250 mg Amber Glass Bottle 250mg
10677703 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10554984 Supplier Thermo Scientific Maybridge Supplier No. MO00686CB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 176445-80-0
Molecular Formula C8H18N2O
Molecular Weight (g/mol) 158.245
MDL Number MFCD09065027
InChI Key KCLFWPREWYWLTP-UHFFFAOYSA-N
Synonym 4-aminomethyl-n,n-dimethyloxan-4-amine, 4-aminomethyl-n,n-dimethyltetrahydro-2h-pyran-4-amine, 4-aminomethyl-tetrahydro-pyran-4-yl-dimethyl-amine, 4-aminomethyl-4-dimethylamino-tetrahydro-4h-pyrane, 2h-pyran-4-methanamine,4-dimethylamino tetrahydro, 4-aminomethyl tetrahydro-2h-pyran-4-yl-dimethylamine, 4-aminomethyl-4-dimethylamino tetrahydro-2h-pyran, 4-aminomethyl 2h-3,4,5,6-tetrahydropyran-4-yl dimethylamine
PubChem CID 10820849
IUPAC Name 4-(aminomethyl)-N,N-dimethyloxan-4-amine
SMILES CN(C)C1(CCOCC1)CN

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 158.24
Percent Purity 95%
Chemical Name or Material 4-(Aminomethyl)-N, N-dimethyltetrahydro-2H-pyran-4-Amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.