missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)indole, 97%

Product Code. 15441098
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15441098 250 mg 250mg
15421098 1 g 1g
15431098 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15441098 Supplier Thermo Scientific Alfa Aesar Supplier No. H54963.MD
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3468-18-6 | C9H10N2 | 146.19 g/mol

4-(Aminomethyl)indole is used as a reactant for preparation of dopamine receptor antagonists and for preparation of high-affinity ligands to α2δ subunit of voltage-gated calcium channels. It is used as a synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Aminomethyl)indole is used as a reactant for preparation of dopamine receptor antagonists and for preparation of high-affinity ligands to α2δ subunit of voltage-gated calcium channels. It is used as a synthetic intermediate.

Solubility
Soluble in ethanol, dimethyl sulfoxide and dimethyl formamide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3468-18-6
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.19
MDL Number MFCD04973297
InChI Key FFBWKPKOXRMLNP-UHFFFAOYSA-N
Synonym 4-aminomethylindole, 1h-indol-4-yl methanamine, 1h-indole-4-methanamine, 4-aminomethyl indole, 1h-indol-4-ylmethyl amine, 1-1h-indol-4-yl methanamine, c-1h-indol-4-yl-methylamine, indole-4-methylamine, 4-aminomethyl-indole, 4-aminomethyl-1h-indole
PubChem CID 280302
IUPAC Name 1H-indol-4-ylmethanamine
SMILES NCC1=C2C=CNC2=CC=C1

Specifications

Melting Point 130°C to 135°C
Quantity 250 mg
Solubility Information Soluble in ethanol,dimethyl sulfoxide and dimethyl formamide.
Formula Weight 146.19
Percent Purity 97%
Chemical Name or Material 4-(Aminomethyl)indole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.