Learn More
Molecular Probes™ 4'-(Aminomethyl)Fluorescein, Hydrochloride
Click to view available options
Quantity:
25 mg
Packaging:
1 Each
Unit Size:
Each
Description
The primary aliphatic amine of 4'-(aminomethyl)fluorescein can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3) to form new probes. Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247).
Specifications
Specifications
| Color | Orange |
| Packaging | 1 Each |
| Quantity | 25 mg |