missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)benzoic acid, MedChemExpress

Product Code. 16422329
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16422329 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16422329 Supplier Medchem Express Supplier No. HYB1258

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

MedChemExpress 4-(Aminomethyl)benzoic acid is an unnatural amino acid derivative, is an antifibrinolytic.

4-(Aminomethyl)benzoic acid is an unnatural amino acid derivative, is an antifibrinolytic.

Specifications

Chemical Name or Material 4-(Aminomethyl)benzoic acid
CAS 56-91-7
Color White
Purity Grade Notes Research
Molecular Formula C8H9NO2
Quantity 1 g
Synonym Aminomethylbenzoic acid α-Amino-p-toluic acid
Solubility Information H2O : 9.09 mg/mL (60.13 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES O=C(O)C1=CC=C(CN)C=C1
Molecular Weight (g/mol) 151.16
Formula Weight 151.16
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.