missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)benzeneboronic acid hydrochloride, 96%

Product Code. 15470156
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15470156 5 g 5g
15460156 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15470156 Supplier Thermo Scientific Alfa Aesar Supplier No. H52855.06
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 75705-21-4 | C7H11BClNO2 | 187.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75705-21-4
Molecular Formula C7H11BClNO2
Molecular Weight (g/mol) 187.43
MDL Number MFCD01632199
InChI Key HUZNRXFJHYNUMV-UHFFFAOYSA-N
Synonym 4-aminomethylphenylboronic acid hydrochloride, 4-aminomethyl phenylboronic acid hydrochloride, 4-aminomethylphenylboronic acid, hcl, 4-aminomethyl benzeneboronic acid hydrochloride, 4-aminomethylphenylboronic acid hcl, 4-aminomethylphenyl boronic acid hydrochloride, 4-aminomethyl phenyl boronic acid hydrochloride, 4-aminomethylbenzeneboronic acid hydrochloride, 4-aminomethylphenylboronic acid, hydrochloride
PubChem CID 2734311
IUPAC Name [4-(aminomethyl)phenyl]boronic acid;hydrochloride
SMILES Cl.NCC1=CC=C(C=C1)B(O)O

Specifications

Melting Point 240°C to 250°C
Quantity 5 g
Formula Weight 187.43
Percent Purity 96%
Chemical Name or Material 4-(Aminomethyl)benzeneboronic acid hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.