missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Aminomethyl)benzeneboronic acid hydrochloride, 96%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 240°C to 250°C |
| Quantity | 1 g |
| Formula Weight | 187.43 |
| Percent Purity | 96% |
| Chemical Name or Material | 4-(Aminomethyl)benzeneboronic acid hydrochloride |
Safety and Handling
Product Identifier
- 4-(Aminomethyl)benzeneboronic acid hydrochloride
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: 4-(Aminomethyl)benzeneboronic acid hydrochloride
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?