missing translation for 'onlineSavingsMsg'
Learn More

4-Aminomethyl-1-Boc-piperidine, 97%

Product Code. p-7035111
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15409587 5 g 5g
15419587 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15409587 Supplier Thermo Scientific Alfa Aesar Supplier No. H52819.06
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 144222-22-0 | C11H23N2O2 | 215.32 g/mol

4-Aminomethyl-1-Boc-piperidine is used in the preparation biologically active compounds and orally active platelet-activating factor antagonists. It is used as an inhibitor for aspartic acid protease and Kinesin spindle protein. It is an active reactant involved in the enantioselective synthesis of N-alkyl terminal aziridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminomethyl-1-Boc-piperidine is used in the preparation biologically active compounds and orally active platelet-activating factor antagonists. It is used as an inhibitor for aspartic acid protease and Kinesin spindle protein. It is an active reactant involved in the enantioselective synthesis of N-alkyl terminal aziridines.

Solubility
Soluble in chloroform and methanol.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144222-22-0
Molecular Formula C11H23N2O2
Molecular Weight (g/mol) 215.32
MDL Number MFCD01076207
InChI Key KLKBCNDBOVRQIJ-UHFFFAOYSA-O
Synonym 1-boc-4-aminomethyl piperidine, tert-butyl 4-aminomethyl piperidine-1-carboxylate, 1-boc-4-aminomethylpiperidine, 1-boc-4-aminomethyl-piperidine, 4-aminomethyl-1-n-boc-piperidine, boc-4-aminomethyl-piperidine, 4-aminomethyl-1-n-t-butoxycarbonyl piperidine, 4-aminomethyl-1-boc-piperidine, 1-n-boc-4-aminomethyl piperidine, 4-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester
PubChem CID 736817
IUPAC Name {1-[(tert-butoxy)carbonyl]piperidin-4-yl}methanaminium
SMILES CC(C)(C)OC(=O)N1CCC(C[NH3+])CC1

Specifications

Density 1.013
Boiling Point 237°C to 238°C
Flash Point >110°C (230°F)
Quantity 5 g
Solubility Information Soluble in chloroform and methanol.
Formula Weight 214.31
Percent Purity 97%
Chemical Name or Material 4-Aminomethyl-1-Boc-piperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.