missing translation for 'onlineSavingsMsg'
Learn More

4-Aminoimidazole-5-carboxamide hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11436926
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11436926 25 g 25g
11416926 1 g 1g
11426926 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11436926 Supplier Thermo Scientific Alfa Aesar Supplier No. B24012.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-40-2
Molecular Formula C4H7ClN4O
Molecular Weight (g/mol) 162.58
MDL Number MFCD00012704
InChI Key MXCUYSMIELHIQL-UHFFFAOYSA-N
Synonym 4-amino-5-imidazolecarboxamide hydrochloride, 5-amino-4-imidazolecarboxamide hydrochloride, 5-amino-1h-imidazole-4-carboxamide hydrochloride, 5-aminoimidazole-4-carboxamide hydrochloride, aic .cntdot. hcl, 4-amino-1h-imidazole-5-carboxamide hydrochloride, aminoimidazolecarboxamide hydrochloride, 4-aminoimidazole-5-carboxamide hydrochloride, 1h-imidazole-4-carboxamide, 5-amino-, monohydrochloride, 5-aminoimidazol-4-carboxamide, hydrochloride
PubChem CID 66146
IUPAC Name 4-amino-1H-imidazole-5-carboxamide;hydrochloride
SMILES Cl.NC(=O)C1=C(N)N=CN1

Specifications

Melting Point ∼270°C (decomposition)
Odor Odorless
Quantity 25 g
Beilstein 3701645
Sensitivity Hygroscopic
Formula Weight 162.58
Percent Purity 98%
Chemical Name or Material 4-Aminoimidazole-5-carboxamide hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.