missing translation for 'onlineSavingsMsg'
Learn More

4-Aminohippuric Acid, TRC

Product Code. 30325269
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325269 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325269 Supplier Toronto Research Chemicals Supplier No. TRCA62797010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aminohippuric Acid
CAS 61-78-9
Molecular Formula C9 H10 N2 O3
Quantity 10 g
Synonym Glycine, N-(4-aminobenzoyl)-, Hippuric acid, p-amino- (8CI), N-(4-Aminobenzoyl)glycine, (4-Aminobenzoylamino)acetic acid, (p-Aminobenzoyl)glycine, 2-[(4-Aminobenzoyl)amino]acetic acid, 4-Aminobenzoylglycine, 4-Aminohippuric acid, N-(4-Aminobenzoyl)aminoacetic acid, N-(p-Aminobenzoyl)glycine, NSC 13064, NSC 7550, PAH, PAH (amino acid), p-Aminohippuric acid
InChI Formula InChI=1S/C9H10N2O3/c10-7-3-1-6(2-4-7)9(14)11-5-8(12)13/h1-4H,5,10H2,(H,11,14)(H,12,13)
SMILES Nc1ccc(cc1)C(=O)NCC(=O)O
IUPAC Name 2-[(4-aminobenzoyl)amino]acetic acid
Molecular Weight (g/mol) 194.19
Formula Weight 194.0691
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.