missing translation for 'onlineSavingsMsg'
Learn More

4-Aminohippuric acid, 99%

Product Code. 10206110 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
10g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10206110 25 g Plastic bottle 25g
10676164 10 g Glass Bottle 10g
10123590 100 g Plastic bottle 100g
10321184 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10206110 Supplier Thermo Scientific Acros Supplier No. 153800250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 61-78-9 | C9H10N2O3 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-78-9
Molecular Weight (g/mol) 194.19
MDL Number MFCD00007890
InChI Key HSMNQINEKMPTIC-UHFFFAOYSA-N
Synonym 4-aminohippuric acid, aminohippuric acid, p-aminohippuric acid, paha, glycine, n-4-aminobenzoyl, n-4-aminobenzoyl glycine, para-aminohippuric acid, aminohippurate, nefrotest, n-p-aminobenzoyl glycine
PubChem CID 2148
ChEBI CHEBI:104011
IUPAC Name 2-[(4-aminobenzoyl)amino]acetic acid
SMILES C1=CC(=CC=C1C(=O)NCC(=O)O)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 198.0°C to 202.0°C
Color Gray to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Molecular Formula C9H10N2O3
Linear Formula H2NC6H4CONHCH2CO2H
Quantity 25 g
Beilstein 14, II, 258
Merck Index 15, 439
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, benzene, aceton and most, organic solvents, pract. insoluble in ccl4, very soluble in dilute hydrochloric acid and alkal
Formula Weight 194.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminohippuric acid
Show More Show Less
Product Identifier
  • 4-Aminohippuric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.