missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobutyric acid, 99+%

Product Code. 15443919
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15443919 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15443919 Supplier Thermo Scientific Alfa Aesar Supplier No. J61307.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56-12-2 | C4H9NO2 | 103.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point ∽195°C (decomposition)
CAS 56-12-2
Color White
Molecular Formula C4H9NO2
Linear Formula H2N(CH2)3CO2H
MDL Number MFCD00008226
Beilstein 906818
Merck Index 14,430
Synonym 4-aminobutyric acid, gaba, gamma-aminobutyric acid, piperidic acid, piperidinic acid, aminalon, gaballon, gammalon, mielogen, butanoic acid, 4-amino
InChI Key BTCSSZJGUNDROE-UHFFFAOYSA-N
SMILES C(CC(=O)O)CN
IUPAC Name 4-aminobutanoic acid
Molecular Weight (g/mol) 103.121
Quantity 10 g
PubChem CID 119
ChEBI CHEBI:16865
Odor Odorless
Formula Weight 103.12
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material 4-Aminobutyric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.