missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobutylcarbamic Acid tert-Butyl Ester, TRC

Product Code. 30309664
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30309664 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309664 Supplier Toronto Research Chemicals Supplier No. TRCA6021402.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Aminobutylcarbamic Acid tert-Butyl Ester
CAS 68076-36-8
Molecular Formula C9H20N2O2
Quantity 2.5 g
Synonym N-(4-Aminobutyl)carbamic Acid 1,1-Dimethylethyl Ester (4-Aminobutyl)carbamic Acid 1,1-Dimethylethyl Ester, 1,1-Dimethylethyl (4-aminobutyl)carbamate, 1-Amino-4-[(tert-butoxycarbonyl)amino]butane, 1-[(tert-Butoxycarbonyl)amino]-4-aminobutane, 4-(tert-Butoxycarbonylamino)butylamine, 4-Aminobutylcarbamic Acid tert-Butyl Ester, 4-Aminobutylcarbamic Acid tert-Butyl Ester, 4-[(tert-Butoxycarbonyl)amino]-1-butylamine, N-(4-Aminobutyl)carbamic Acid tert-Butyl Ester, N-(4-Aminobutyl)carbamic Acid tert-Butyl Ester, N-(tert-Butoxycarbonyl)-1,4-Butanediamine, N-(tert-Butoxycarbonyl)putrescine, N-Boc-butane-1,4-Diamine, N-tert-Butoxycarbonyl-1,4-diaminobutane, tert-Butyl 4-Aminobutylcarbamate, tert-Butyl N-(4-aminobutyl)carbamate
InChI Formula InChI=1S/C9H20N2O2/c1-9(2,3)13-8(12)11-7-5-4-6-10/h4-7,10H2,1-3H3,(H,11,12)
SMILES CC(C)(C)OC(=O)NCCCCN
IUPAC Name tert-butyl N-(4-aminobutyl)carbamate
Molecular Weight (g/mol) 188.27
Formula Weight 188.1525
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.