missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobutanoic Acid (gamma-Aminobutyric Acid), Mikromol™

Product Code. 30051946
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051946 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051946 Supplier Mikromol Supplier No. MM1296.010100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Aminobutanoic Acid (gamma-Aminobutyric Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Aminobutanoic acid (GABA)
CAS 56-12-2
Analyte or Component Names 4-Aminobutanoic acid (GABA)
Shipping Condition Room Temperature
Molecular Formula C4 H9 N O2
Certifications/Compliance ISO 9001
Synonym Vigabatrin impurity D (EP), Gamma Aminobutyric Acid, 4-Aminobutanoic Acid, γ-Aminobutyric Acid, Butanoic acid, 4-amino-, Butyric acid, 4-amino- (7CI,8CI), 3-Carboxypropylamine, 4-Aminobutyric acid, Aminalon, GABA, Gaballon, Gamarex, Gammalon, Gammalone, Gammar, Gammasol, Mielogen, Mielomade, NSC 27418, NSC 32044, NSC 45460, NSC 51295, Oryza GABA Extract HC 90, Pharma-GABA 20S, Pharmagaba, Pharmagaba 20, Pharmagaba 20D, Piperidic acid, Piperidinic acid, γ-Aminobutanoic acid, ω-Aminobutyric acid, Sodium Alendronate Impurity A, Vigabatrin Impurity D
InChI Formula InChI=1S/C4H9NO2/c5-3-1-2-4(6)7/h1-3,5H2,(H,6,7)
SMILES NCCCC(=O)O
IUPAC Name 4-aminobutanoic acid
Molecular Weight (g/mol) 103.12
Quantity 100 mg
Formula Weight 103.0633 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.