missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobiphenyl, TRC

Product Code. 17939289
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17939289 1 g 1g
17997079 5 g 5g
17928899 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 17939289 Supplier Toronto Research Chemicals Supplier No. TRCA6017811G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-67-1
Molecular Formula C12 H11 N
Molecular Weight (g/mol) 169.22
Synonym 4-Biphenylamine (8CI), (1,1'-Biphenyl-4-yl)amine, 4-Amino-1,1'-biphenyl, 4-Aminobiphenyl, 4-Aminodiphenyl, 4-Biphenylylamine, 4-Phenylaniline, 4-Phenylbenzenamine, NSC 7660, Xenylamine, p-Aminobiphenyl, p-Aminodiphenyl, p-Aminophenyl benzene, p-Biphenylamine, p-Phenylaniline, p-Xenylamine
IUPAC Name 4-phenylaniline
SMILES Nc1ccc(cc1)c2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H11N/c13-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9H,13H2
Formula Weight 169.0891
Chemical Name or Material 4-Aminobiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.