missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzyl alcohol, 98%

Product Code. p-7021692
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448096 5 g 5g
11458096 25 g 25g
11468096 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448096 Supplier Thermo Scientific Alfa Aesar Supplier No. A14130.06
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 623-04-1 | C7H9NO | 123.16 g/mol

4-Aminobenzyl alcohol is used in the synthesis of 4-{N-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutylamino}benzyl ester. It is also used as an intermediate in organic synthesis and pharmaceutical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzyl alcohol is used in the synthesis of 4-{N-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutylamino}benzyl ester. It is also used as an intermediate in organic synthesis and pharmaceutical products.

Solubility
Soluble in alcohol, ether and benzene. Partially soluble in water.

Notes
Air and light sensitive. Incompatible with strong oxidizing agents and strong acids. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-04-1
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00014782
InChI Key AXKGIPZJYUNAIW-UHFFFAOYSA-N
Synonym 4-aminobenzyl alcohol, 4-aminophenyl methanol, 4-aminobenzylalcohol, 4-hydroxymethyl aniline, p-aminobenzyl alcohol, p-aminobenzylalcohol, benzenemethanol, 4-amino, benzyl alcohol, p-amino, 4-amino-benzylalcohol, pubchem7480
PubChem CID 69331
IUPAC Name (4-aminophenyl)methanol
SMILES NC1=CC=C(CO)C=C1

Specifications

Melting Point 59°C to 63°C
Quantity 5 g
Beilstein 2078680
Sensitivity Air and light sensitive
Solubility Information Soluble in alcohol,ether and benzene. Partially soluble in water.
Formula Weight 123.16
Percent Purity 98%
Chemical Name or Material 4-Aminobenzyl alcohol
compliance-icons
Product Identifier
  • 4-Aminobenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-Aminobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.