missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzophenone, 98%

Product Code. 11414804
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414804 10 g 10g
11424804 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414804 Supplier Thermo Scientific Alfa Aesar Supplier No. A13757.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1137-41-3 | C13H11NO | 197.24 g/mol

4-Aminobenzophenone is associated with 4-benzoylpyridine and it is used as a frequency doubler for semiconductor lasers. It is used in the preparation of 1-[4-[N,N-bis(trimethylsilyl)amino]phenyl]-1-phenylethylene. It is used as a precursor to prepare 1,2,3-triazoles, pyrimidotriazoles, benzothiazole and pyrimidinone derivatives. It is involved in the formation of coordination complexes with chromium(III).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzophenone is associated with 4-benzoylpyridine and it is used as a frequency doubler for semiconductor lasers. It is used in the preparation of 1-[4-[N,N-bis(trimethylsilyl)amino]phenyl]-1-phenylethylene. It is used as a precursor to prepare 1,2,3-triazoles, pyrimidotriazoles, benzothiazole and pyrimidinone derivatives. It is involved in the formation of coordination complexes with chromium(III).

Solubility
Soluble in cold water, alcohol and ether.

Notes
Air and light sensitive. Incompatible with strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1137-41-3
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.24
MDL Number MFCD00007895
InChI Key RBKHNGHPZZZJCI-UHFFFAOYSA-N
Synonym 4-aminobenzophenone, 4-aminophenyl phenyl methanone, 4-benzoylaniline, p-aminobenzophenone, methanone, 4-aminophenyl phenyl, benzophenone, 4-amino, p-benzoylaniline, usaf a-233, 4-aminophenyl-phenylmethanone, 4-aminophenyl-phenyl-methanone
PubChem CID 14346
IUPAC Name (4-aminophenyl)-phenylmethanone
SMILES NC1=CC=C(C=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 121°C to 124°C
Boiling Point 245°C to 247°C (22 mmHg)
Quantity 10 g
Beilstein 389292
Solubility Information Soluble in cold water,alcohol and ether.
Formula Weight 197.24
Percent Purity 98%
Chemical Name or Material 4-Aminobenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.