missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzonitrile, 98%

Product Code. 11494864
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494864 50 g 50g
11484864 10 g 10g
11404874 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494864 Supplier Thermo Scientific Alfa Aesar Supplier No. A13794.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 873-74-5 | C7H6N2 | 118.139 g/mol

4-Aminobenzonitrile is used as an amino substituted benzonitrile with hypotensive activity and as a radioprotective agent. It is also used as a derivatization reagent in the capillary zone electrophoretic analysis of aldoses, ketoses and uronic acid. Further, it is used in the preparation of methacrylic monomers containing pendant azobenzene structures and polythiophenes containing an azobenzene moiety in the side-chain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzonitrile is used as an amino substituted benzonitrile with hypotensive activity and as a radioprotective agent. It is also used as a derivatization reagent in the capillary zone electrophoretic analysis of aldoses, ketoses and uronic acid. Further, it is used in the preparation of methacrylic monomers containing pendant azobenzene structures and polythiophenes containing an azobenzene moiety in the side-chain.

Solubility
Soluble in ethyl acetate, dichloromethane and chloroform. Insoluble in water.

Notes
Light sensitive. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-74-5
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00007821
InChI Key YBAZINRZQSAIAY-UHFFFAOYSA-N
Synonym 4-cyanoaniline, p-aminobenzonitrile, p-cyanoaniline, benzonitrile, 4-amino, aniline, p-cyano, benzonitrile, p-amino, 1-amino-4-cyanobenzene, unii-pb8v69y76t, 4-amino benzonitrile, 4-amino-benzonitrile
PubChem CID 13396
IUPAC Name 4-aminobenzonitrile
SMILES C1=CC(=CC=C1C#N)N

Specifications

Melting Point 83°C to 87°C
Quantity 50 g
UN Number UN3439
Beilstein 774507
Solubility Information Soluble in ethyl acetate,dichloromethane and chloroform. Insoluble in water.
Formula Weight 118.14
Percent Purity 98%
Chemical Name or Material 4-Aminobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.