missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzoic acid sodium salt, 98%

Product Code. 15415679
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15415679 25 g 25g
15425679 100 g 100g
15435679 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15415679 Supplier Thermo Scientific Alfa Aesar Supplier No. J63428.14
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

Pharmaceutical intermediate | CAS: 555-06-6 | C7H6NNaO2 | 159.12 g/mol

4-Aminobenzoic acid sodium salt is used as a Pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzoic acid sodium salt is used as a Pharmaceutical intermediate.

Solubility
Soluble in water (50 mg/ml).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-06-6
Molecular Formula C7H6NNaO2
Molecular Weight (g/mol) 159.12
MDL Number MFCD00064395
InChI Key XETSAYZRDCRPJY-UHFFFAOYSA-M
Synonym sodium 4-aminobenzoate, sodium p-aminobenzoate, 4-aminobenzoic acid sodium salt, sodium aminobenzoate, aminobenzoate sodium, antergyl, pabavit, benzoic acid, 4-amino-, monosodium salt, monosodium 4-aminobenzoate, unii-75ui7quz5j
PubChem CID 517441
IUPAC Name sodium;4-aminobenzoate
SMILES C1=CC(=CC=C1C(=O)[O-])N.[Na+]

Specifications

Melting Point >300°C
Color Yellow
Quantity 25 g
Solubility Information Soluble in water (50mg/ml).
Formula Weight 159.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Aminobenzoic acid sodium salt
compliance-icons
Product Identifier
  • 4-Aminobenzoic acid sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 4-Aminobenzoic acidsodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.