missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzoic Acid, Mikromol™

Product Code. 17987924
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17987924 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17987924 Supplier Mikromol Supplier No. MM0094.03
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Aminobenzoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vitamin H1 (4-Aminobenzoic acid)
CAS 150-13-0
Analyte or Component Names Vitamin H1 (4-Aminobenzoic acid)
Shipping Condition Room Temperature
Molecular Formula C7 H7 N O2
Certifications/Compliance ISO 17034
Synonym Benzylpenicillin Procaine Imp. A (EP), Oxybuprocaine Hydrochloride Imp. A (EP), Oxybuprocaine Imp. A (EP), Tetracaine Hydrochloride Imp. A (EP), Tetracaine Imp. A (EP), 4-Aminobenzoic Acid, 4-Carboxyaniline, 4-Carboxyphenylamine, Actipol, Amben, Aniline-4-carboxylic acid, Anti-Chromotrichia factor, Anticanitic vitamin, Anticantic vitamin, Antichromotrichia factor, Bacterial vitamin H1, Chromotrichia factor, Hachemina, NSC 7627, PAB, PABA, Pabacyd, Pabafilm, Pabamine, Paraminol, Paranate, Romavit, Sunbrella, Trichochromogenic factor, Vitamin BX, Vitamin H', p-Aminobenzoic acid, p-Carboxyaniline, p-Carboxyphenylamine, Benzocaine Impurity G, Benzylpenicillin Procaine Impurity A, Oxybuprocaine Hydrochloride Impurity A, Tetracaine Hydrochloride Impurity A, Oxybuprocaine Impurity A, Tetracaine Impurity A
InChI Formula InChI=1S/C7H7NO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,8H2,(H,9,10)
SMILES Nc1ccc(cc1)C(=O)O
IUPAC Name 4-aminobenzoic acid
Molecular Weight (g/mol) 137.14
Quantity 100 mg
Formula Weight 137.0477 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.