missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzoic Acid Methyl Ester, TRC

Product Code. 30314340
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314340 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314340 Supplier Toronto Research Chemicals Supplier No. TRCA59150525G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 4-Aminobenzoate
CAS 619-45-4
Molecular Formula C8 H9 N O2
Quantity 25 g
Synonym Benzoic acid, 4-amino-, methyl ester, Benzoic acid, p-amino-, methyl ester (6CI,7CI,8CI), (4-(Methoxycarbonyl)phenyl)amine, 4-(Carbomethoxy)aniline, 4-(Methoxycarbonyl)aniline, 4-Aminobenzenecarboxylic acid methyl ester, 4-Aminobenzoic acid methyl ester, Methyl 4-aminobenzoate, Methyl 4-aminophenylcarboxylate, Methyl aniline-4-carboxylate, Methyl p-aminobenzoate, NSC 3783, p-(Methoxycarbonyl)aniline, p-Aminobenzoic acid methyl ester, p-Carbomethoxyaniline
InChI Formula InChI=1S/C8H9NO2/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5H,9H2,1H3
SMILES COC(=O)c1ccc(N)cc1
IUPAC Name methyl 4-aminobenzoate
Molecular Weight (g/mol) 151.16
Formula Weight 151.0633
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.