missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzoic acid, MedChemExpress

Product Code. 16491088
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491088 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16491088 Supplier Medchem Express Supplier No. HYB1008
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 4-Aminobenzoic acid is an intermediate in the synthesis of folate by bacteria, plants, and fungi.

4-Aminobenzoic acid is an intermediate in the synthesis of folate by bacteria, plants, and fungi.

Specifications

Chemical Name or Material 4-Aminobenzoic acid
Physical Form Solid
CAS 150-13-0
Color Off-White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C7H7NO2
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym PABA Vitamin Bx Vitamin H1 p-Aminobenzoic acid
Solubility Information DMSO : ≥ 100 mg/mL (729.18 mM) ∣H2O : 4.55 mg/mL (33.18 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 137.14
Formula Weight 137.14
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.