missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzenesulphonamide (Sulphanilamide), Mikromol™

Product Code. 17913416
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17913416 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17913416 Supplier Mikromol Supplier No. MM0227.050250
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Aminobenzenesulphonamide (Sulphanilamide)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfanilamide
CAS 63-74-1
Analyte or Component Names Sulfanilamide
Shipping Condition Room Temperature
Molecular Formula C6 H8 N2 O2 S
Certifications/Compliance ISO 17034
Synonym Sulfacetamide Sodium Imp. A (EP), Sulfacetamide Imp. A (EP), Sulfadiazine Imp. D (EP), Sulfadimidine Imp. D (EP), Sulfaguanidine Imp. A (EP), Sulfamethoxazole Imp. E (EP), 4-Aminobenzenesulfonamide, Sulfanilamide, 4-Aminobenzenesulphonamide, Sulphanilamide, Sulfadimidine Imp. D (Pharmeuropa), Sulfadiazine Imp. D (Pharmeuropa), (4-(Aminosulfonyl)phenyl)amine, 1162F, 4-(Aminosulfonyl)aniline, 4-Aminophenylsulfonamide, 4-Sulfamoylaniline, A-349, Albexan, Albosal, Ambeside, Antistrept, Astreptine, Astrocid, Bacteramid, Bactesid, Collomide, Colsulanyde, Copticide, Deseptyl, Dipron, Ergaseptine, Erysipan, Estreptocida, Exoseptoplix, F 1162, Fourneau 1162, Gerison, Gombardol, Infepan, Lusil, Lysococcine, NSC 7618, Neococcyl, Orgaseptine, PABS, Prontalbin, Prontosil Album, Prontosil I, Prontosil white, Prontylin, Pronzin Album, Proseptal, Proseptine, Proseptol, Pysococcine, Ro 1-3354, Rubiazol A, Sanamid, Septamide Album, Septanilam, Septinal, Septolix, Septoplex, Septoplix, Stopton Album, Stramid, Strepamide, Strepsan, Streptagol, Streptamid, Streptasol, Streptocid, Streptocid album, Streptocide, Streptocide (white), Streptoclase, Streptocom, Strepton, Streptopan, Streptosil, Streptozol, Streptozone, Sulfamidyl, Sulfamine, Sulfana, Sulfanalone, Sulfanidyl, Sulfanil, Sulfanilamide Vaginal Cream, Sulfocidine, Sulfonamide, Sulfonamide P, Sulfonylamide, Sulphonamide, Therapol, Tolder, White streptocide, p-Aminobenzenesulfonamide, p-Aminophenylsulfonamide, p-Anilinesulfonamide, p-Sulfamidoaniline, p-Sulfamoylaniline, Ph Eur Sulfamethoxazole Impurity E, Sulfanilamide, Sulfacetamide Sodium Impurity A, Sulfadiazine Impurity D, Sulfadimethoxine Impurity E, Sulfadimethoxine Sodium Impurity E, Sulfadimidine Impurity D, Sulfaguanidine Impurity A, Sulfamethoxazole Impurity E, Sulfacetamide Impurity A, Sulfadimethoxine Impurity E
InChI Formula InChI=1S/C6H8N2O2S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H2,8,9,10)
SMILES Nc1ccc(cc1)S(=O)(=O)N
IUPAC Name 4-aminobenzenesulfonamide
Molecular Weight (g/mol) 172.2
Quantity 250 mg
Formula Weight 172.0306 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.