missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzeneboronic acid pinacol ester, 98%

Product Code. 15408137
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15408137 1 g 1g
15418137 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15408137 Supplier Thermo Scientific Alfa Aesar Supplier No. H51666.03
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 214360-73-3 | C12H18BNO2 | 219.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Aminobenzeneboronic acid pinacol ester is a reagent used for Suzuki-Miyaura cross-coupling reactions, Mercury(II) detection by fluorometry with new fluorogenic indicators based on through-bond energy transfer from pentaquinone to rhodamine, Rhodium-catalyzed amination reactions and Palladium-catalyzed Suzuki cross-coupling to synthesize potential antitubercular and antimicrobial compounds. It is also used as a reagent in preparation of Hexaphenylbenzene derivatives as a potential bioprobe and multichannel keypad system, Pyromellitic diimide-based polymer as matrix for solution-processable n-channel field-effect transistors, alternating copolymers of oligoarylenes and naphthalene bisimides as low band gap semiconductors with electrochemical and spectroelectrochemical behavior and γ-secretase modulators in the treatment of Amyloid β formation.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 214360-73-3
Molecular Formula C12H18BNO2
Molecular Weight (g/mol) 219.09
MDL Number MFCD02093721
InChI Key ZANPJXNYBVVNSD-UHFFFAOYSA-N
Synonym 4-aminophenylboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-aminophenylboronic acid pinacolate, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, pinacol cyclic ester, 2-4-aminophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-aminophenylboronic acid, pinacol ester, 4-aminophenylboronicacidpinacolester, 4-aminobenzeneboronic acid pinacol ester
PubChem CID 2734620
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(N)C=C1

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 219.09
Percent Purity 98%
Chemical Name or Material 4-Aminobenzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.