missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzeneboronic acid hydrochloride, 97%

Product Code. 11387359
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387359 500 mg 500mg
11377359 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387359 Supplier Thermo Scientific Alfa Aesar Supplier No. H27479.ME
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 80460-73-7 | C6H9BClNO2 | 173.403 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80460-73-7
Molecular Formula C6H9BClNO2
Molecular Weight (g/mol) 173.403
MDL Number MFCD03001333
InChI Key QBYGJJSFMOVYOA-UHFFFAOYSA-N
Synonym 4-aminophenylboronic acid hydrochloride, 4-aminophenyl boronic acid hydrochloride, 4-aminobenzeneboronic acid hydrochloride, 4-aminophenylboronic acid hcl, 4-boronoaniline hydrochloride, 4-aminophenylboronic acid, hcl, boronic acid, 4-aminophenyl-, hydrochloride, pubchem1747, 4-aminophenylboronicacidhydrochloride, ksc494e4p
PubChem CID 2734614
IUPAC Name (4-aminophenyl)boronic acid;hydrochloride
SMILES B(C1=CC=C(C=C1)N)(O)O.Cl

Specifications

Melting Point 125°C to 132°C
Quantity 500 mg
Sensitivity Hygroscopic
Formula Weight 173.41
Percent Purity 97%
Chemical Name or Material 4-Aminobenzeneboronic acid hydrochloride
compliance-icons
Product Identifier
  • 4-Aminobenzeneboronic acid hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-Aminobenzeneboronic acid hydrochloride

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.