missing translation for 'onlineSavingsMsg'
Learn More

4'-Aminobenzanilide, 97%

Product Code. 11481677
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481677 25 g 25g
11471677 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11481677 Supplier Thermo Scientific Alfa Aesar Supplier No. B25663.14
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 17625-83-1 | C13H12N2O | 212.252 g/mol

4'-Aminobenzanilide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field. It has anticonvulsant property.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Aminobenzanilide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field. It has anticonvulsant property.

Solubility
Soluble in DMSO and methanol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17625-83-1
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.252
MDL Number MFCD00035777
InChI Key GTTFJYUWPUKXJH-UHFFFAOYSA-N
Synonym 4'-aminobenzanilide, n-4-aminophenyl benzamide, benzamide, n-4-aminophenyl, 4,4'-diaminobenzoylanilide, n-4-amino-phenyl-benzamide, n-4-aminophenyl-benzamide, 4-aminobenzanilide, 4/'-aminobenzanilide, acmc-209xfr, n-benzoyl 4-aminoaniline
PubChem CID 87196
IUPAC Name N-(4-aminophenyl)benzamide
SMILES C1=CC=C(C=C1)C(=O)NC2=CC=C(C=C2)N

Specifications

Melting Point 127°C to 131°C
Quantity 25 g
Solubility Information Soluble in DMSO and methanol.
Formula Weight 212.25
Percent Purity 97%
Chemical Name or Material 4'-Aminobenzanilide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.