missing translation for 'onlineSavingsMsg'
Learn More

4-Aminoantipyrine, 99%, MP Biomedicals™

Product Code. 11493141 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
250 g
Unit Size:
100g
10g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493141 100 g 100g
11483141 10 g 10g
11403151 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493141 Supplier MP Biomedicals Supplier No. 0215034380
 Restricted Item
Estimated Shipment: 08-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

4-Aminoantipyrine, Yellow Powder

  • Reagent for determination of chlorinated phenols, phenylglycosidase, and glucose in the presence of phenol and peroxidase.

Chemical Identifiers

CAS 83-07-8
Molecular Formula C11H13N3O
MDL Number MFCD00003145
InChI Key RLFWWDJHLFCNIJ-UHFFFAOYSA-N
PubChem CID 2151
ChEBI CHEBI:59026
IUPAC Name 4-amino-1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CC1=C(C(=O)N(N1C)C2=CC=CC=C2)N

Specifications

Color Yellow
Assay Percent Range 0.99
Quantity 100 g
Synonym 4-aminoantipyrine, ampyrone, metapirazone, aminoantipyrine, solvapyrin-a, aminoazophene, 4-aminophenazone, 4-aminoantipyrene, aminoantipyrin, solnapyrin-a
Molecular Weight (g/mol) 203.245
Formula Weight 203.25
Physical Form Powder
Chemical Name or Material 4-Aminoantipyrine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.