missing translation for 'onlineSavingsMsg'
Learn More

4'-Aminoacetanilide, 95%

Product Code. 10045720 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10045720 100 g Glass bottle 100g
10219550 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10045720 Supplier Thermo Scientific Acros Supplier No. 400601000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122-80-5 | C8H10N2O | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-80-5
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.18
MDL Number MFCD00007853
InChI Key CHMBIJAOCISYEW-UHFFFAOYSA-N
Synonym 4'-aminoacetanilide, n-4-aminophenyl acetamide, p-aminoacetanilide, 4-aminoacetanilide, 4-acetamidoaniline, p-amino acetanilide, p-acetamidoaniline, acetparamin, acetamide, n-4-aminophenyl, fourrine a
PubChem CID 31230
IUPAC Name N-(4-aminophenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 159.0°C to 165.0°C
Color Brown to Pink
Boiling Point 267.0°C
Flash Point >150°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula NHCOCH3C6H4NH2
Quantity 100 g
Merck Index 15, 405
Solubility Information Solubility in water: soluble in cold water. Other solubilities: freely soluble in hot water,freely soluble in alcohol,ether
Formula Weight 150.18
Percent Purity 95%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4'-Aminoacetanilide, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Aminoacetanilide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.