missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-TEMPO, ≥98.0% (GC), Honeywell™ Fluka™

Product Code. 15605540 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15605540 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15605540 Supplier Honeywell-Fluka Supplier No. 43967250MG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 14691-88-4
Color Red
Packaging Glass bottle
MDL Number MFCD00006479
Quantity 250 mg
UN Number NONH for all modes of transport
Beilstein 3933966
Formula Weight 171.26g/mol
Percent Purity ≥98.0% (GC)
Physical Form Solid
Chemical Name or Material 4-Amino-TEMPO
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.