missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-L-phenylalanine, 95%

Product Code. 15428437
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15428437 5 g 5g
15438437 250 mg 250mg
15418437 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15428437 Supplier Thermo Scientific Alfa Aesar Supplier No. H51970.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 943-80-6 | C9H12N2O2 | 180.21 g/mol

It finds its application in the purification of peptide synthetases involved in pristinamycin I biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the purification of peptide synthetases involved in pristinamycin I biosynthesis.

Solubility
Soluble in water.

Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 943-80-6
Molecular Formula C9H12N2O2
Molecular Weight (g/mol) 180.21
MDL Number MFCD00069927
InChI Key CMUHFUGDYMFHEI-QMMMGPOBSA-N
Synonym 4-amino-l-phenylalanine, h-phe 4-nh2-oh, 4-aminophenylalanine, l-4-aminophenylalanine, p-amino-l-phenylalanine, p-aminophenylalanine, s-2-amino-3-4-aminophenyl propanoic acid, 4-amino-phenylalanine, 2s-2-amino-3-4-aminophenyl propanoic acid, l-4-nh2-phe-oh
PubChem CID 151001
ChEBI CHEBI:29737
IUPAC Name (2S)-2-amino-3-(4-aminophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(N)C=C1)C(O)=O

Specifications

Solubility Information Soluble in water.
Quantity 5 g
Formula Weight 180.2
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material 4-Amino-L-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.