missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-hydroxypyrimidine, 98%, Thermo Scientific™

Product Code. 15519503 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15519503 5 g 5g
15509503 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15519503 Supplier Thermo Scientific Acros Supplier No. 459750050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1193-22-2
Molecular Formula C4H5N3O
Molecular Weight (g/mol) 111.10
MDL Number MFCD00051502
InChI Key HFMLLTVIMFEQRE-UHFFFAOYSA-N
Synonym 4-amino-6-hydroxypyrimidine, 6-aminopyrimidin-4-ol, 4-hydroxy-6-aminopyrimidine, 4-pyrimidinol, 6-amino, 4 1h-pyrimidinone, 6-amino, 6-aminopyrimidin-4 3h-one, 4-oxy-6-aminopyrimidine, 6-amino-4-pyrimidinol, 6-amino-3h-pyrimidin-4-one, 6-amino-4 1h-pyrimidinone
PubChem CID 70944
SMILES NC1=CC(=O)N=CN1

Specifications

Melting Point 262.0°C to 264.0°C
Loss on Drying 1% max. (105°C, 3 hr)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: slightly soluble.
IUPAC Name 6-amino-1,4-dihydropyrimidin-4-one
Formula Weight 111.1
Percent Purity 98%
Chemical Name or Material 4-Amino-6-hydroxypyrimidine
compliance-icons
Product Identifier
  • 4-Amino-6-hydroxypyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.