missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-hydroxy-2-mercaptopyrimidine monohydrate, 98+%

Product Code. 11361467
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361467 25 g 25g
11371467 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361467 Supplier Thermo Scientific Alfa Aesar Supplier No. L01992.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 65802-56-4 | C4H7N3O2S | 161.179 g/mol

4-Amino-6-hydroxy-2-mercaptopyrimidine is a novel, potent, and selective A3 adenosine receptor antagonists. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-6-hydroxy-2-mercaptopyrimidine is a novel, potent, and selective A3 adenosine receptor antagonists. It is also used as pharmaceutical intermediates.

Solubility
Soluble in water and DMSO.

Notes
Store at 4°C. Incompatible with oxidizing agents. Avoid excessive heat and light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65802-56-4
Molecular Formula C4H7N3O2S
Molecular Weight (g/mol) 161.179
MDL Number MFCD00150556
InChI Key BWYGMIYEADAIGW-UHFFFAOYSA-N
Synonym 4-amino-6-hydroxy-2-mercaptopyrimidine monohydrate, 6-amino-2-thiouracil monohydrate, 6-amino-2-thioxo-2,3-dihydropyrimidin-4 1h-one hydrate, 2-pyrimidinethiol, 4-amino-6-hydroxy-, monohydrate, 4-amino-6-hydroxy-2-mercaptopyrimidine hydrate, 6-amino-2-sulfanylpyrimidin-4-ol hydrate, 6-amino-2-mercaptopyrimidin-4-ol hydrate, 4 1h-pyrimidinone, 6-amino-2,3-dihydro-2-thioxo-, monohydrate, 6-amino-2-mercapto-4-pyrimidinol monohydrate, 6-amino-2-sulfanylidene-1h-pyrimidin-4-one hydrate
PubChem CID 2723599
IUPAC Name 6-amino-2-sulfanylidene-1H-pyrimidin-4-one;hydrate
SMILES C1=C(NC(=S)NC1=O)N.O

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 5795441
Solubility Information Soluble in water and DMSO.
Formula Weight 161.19 (143.17 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material 4-Amino-6-hydroxy-2-mercaptopyrimidine monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.