missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5-imidazolecarboxamide hydrochloride, 98%

Product Code. 10405295 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10405295 1 g Glass bottle 1g
10275940 5 g Glass bottle 5g
10684852 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10405295 Supplier Thermo Scientific Acros Supplier No. 173090010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 72-40-2 | C4H7ClN4O | 162.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-40-2 , 360-97-4
Molecular Formula C4H7ClN4O
Molecular Weight (g/mol) 162.58
MDL Number MFCD00012704
InChI Key MXCUYSMIELHIQL-UHFFFAOYSA-N
Synonym 4-amino-5-imidazolecarboxamide hydrochloride, 5-amino-4-imidazolecarboxamide hydrochloride, 5-amino-1h-imidazole-4-carboxamide hydrochloride, 5-aminoimidazole-4-carboxamide hydrochloride, aic .cntdot. hcl, 4-amino-1h-imidazole-5-carboxamide hydrochloride, aminoimidazolecarboxamide hydrochloride, 4-aminoimidazole-5-carboxamide hydrochloride, 1h-imidazole-4-carboxamide, 5-amino-, monohydrochloride, 5-aminoimidazol-4-carboxamide, hydrochloride
PubChem CID 66146
IUPAC Name 4-amino-1H-imidazole-5-carboxamide;hydrochloride
SMILES Cl.NC(=O)C1=C(N)N=CN1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 250°C
Color White
Ignition Residue 0.2% max.
Loss on Drying 0.5% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Quantity 1 g
Beilstein 25, II, 221
Solubility Information (5% in water) Clear
Formula Weight 162.58
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Amino-5-imidazolecarboxamide hydrochloride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.