missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5-cyano-2-methylpyrimidine, 95%, Thermo Scientific™

Product Code. 15417878
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Amino-5-cyano-2-methylpyrimidine is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water (6.4 g/L at 25°C). Soluble in dimethyl sulfoxide, ethanol and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 698-29-3
Molecular Formula C6H6N4
Molecular Weight (g/mol) 134.14
MDL Number MFCD00084875
InChI Key YBPNIILOUYAGIF-UHFFFAOYSA-N
Synonym 4-amino-5-cyano-2-methylpyrimidine, 5-pyrimidinecarbonitrile, 4-amino-2-methyl, 4-amino-2-methyl-5-pyrimidinecarbonitrile, pynitrile, pubchem6987, 2-methyl-4-amino-5-cyan-pyrimidin, 4-amino-5-cyano-2-methyl-pyrimidine, 2-methyl-4-amino-5-cyanopyrimidine, 4-amino-2-methyl-5-pyrimidine carbonitrile, 4-amino-2-methyl-pyrimidine-5-carbonitrile
PubChem CID 69682
IUPAC Name 4-amino-2-methylpyrimidine-5-carbonitrile
SMILES CC1=NC=C(C#N)C(N)=N1

Specifications

Color Yellow
Quantity 250 mg
UN Number UN3439
Solubility Information Slightly soluble in water (6.4g/L at 25°C). Soluble in dimethyl sulfoxide,ethanol and methanol.
Formula Weight 134.14
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-Amino-5-cyano-2-methylpyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'