missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-5-bromo-2-chloropyrimidine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Especificaciones
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 189.0°C to 191.0°C |
Color | Brown to Purple |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Quantity | 1 g |
Formula Weight | 208.45 |
Percent Purity | 97% |
Mostrar más |
Seguridad y manipulación
Nombre del producto
- 4-Amino-5-bromo-2-chloropyrimidine
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'