missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5-aminomethyl-2-methylpyrimidine Dihydrochloride, TRC

Product Code. 30008946
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30008946 50 mg 50mg
30006912 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30008946 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCA58187550MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874-43-1
Molecular Formula C6 H10 N4 . 2 Cl H
Molecular Weight (g/mol) 211.09
Synonym Thiamine Impurity 6 Dihydrochloride
IUPAC Name 5-(aminomethyl)-2-methylpyrimidin-4-amine;dihydrochloride
SMILES Cl.Cl.Cc1ncc(CN)c(N)n1

Specifications

Analyte or Component Names 4-Amino-5-aminomethyl-2-methylpyrimidine, Dihydrochloride
Shipping Condition Room Temperature
InChI Formula InChI=1S/C6H10N4.2ClH/c1-4-9-3-5(2-7)6(8)10-4;;/h3H,2,7H2,1H3,(H2,8,9,10);2*1H
Solution Type Neat
Industry Type Forensics
Quantity 50 mg
Formula Weight 210.0439
Chemical Name or Material 4-Amino-5-aminomethyl-2-methylpyrimidine, Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.