missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5,6-dichloropyrimidine, 98%, Thermo Scientific™

Product Code. 15481999
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15481999 5 g 5g
15471999 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15481999 Supplier Thermo Scientific Alfa Aesar Supplier No. H64691.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 310400-38-5
Molecular Formula C4H3Cl2N3
Molecular Weight (g/mol) 163.99
MDL Number MFCD09837293
InChI Key ISTJUVRUUBNZNB-UHFFFAOYSA-N
Synonym 4-amino-5,6-dichloropyrimidine, 5,6-dichloro-pyrimidin-4-ylamine, 4-pyrimidinamine, 5,6-dichloro, 5,6-dichloro-4-pyrimidinamine, 5,6-dichloropyrimidin-4-ylamine, 4-pyrimidinamine, 5,6-dichloro-9ci
PubChem CID 24219262
IUPAC Name 5,6-dichloropyrimidin-4-amine
SMILES NC1=C(Cl)C(Cl)=NC=N1

Specifications

Color White
Quantity 5 g
Beilstein 8678505
Solubility Information Slightly soluble in water.
Formula Weight 163.99
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Amino-5,6-dichloropyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.