missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-4H-1,2,4-triazole, 99%

Product Code. p-3756849 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10365780

Brand: Thermo Scientific Acros 104700250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 584-13-4 | C2H4N4 | 84.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-13-4
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.08
MDL Number MFCD00003099
InChI Key FMCUPJKTGNBGEC-UHFFFAOYSA-N
Synonym 4-amino-1,2,4-triazole, 4-amino-4h-1,2,4-triazole, 4h-1,2,4-triazol-4-amine, 1-amino-1,3,4-triazole, 4h-1,2,4-triazole, 4-amino, 1-amino-1h-1,3,4-triazole, 4-amino-1,2,4 4h-triazole, 4h-1,2,4-triazol-4-ylamine, 1-amino-1,4-triazole, 4-amino-1,4-triazole
PubChem CID 11432
IUPAC Name 1,2,4-triazol-4-amine
SMILES C1=NN=CN1N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 84.0°C
Color Beige or White
Flash Point >149°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 26, IV, 40
Solubility Information Solubility in water: 810g/L (20°C) in water. Other solubilities: soluble in ethanol and dimethyl sulfoxyde,very little soluble in vegetal oils
Formula Weight 84.08
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-4H-1, 2, 4-triazole
Show More Show Less
compliance-icons
Product Identifier
  • 4-Amino-4H-1,2,4-triazole
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.