missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Product Code. 11308946
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11308946 100 mg 100mg
11398936 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11308946 Supplier Thermo Scientific Alfa Aesar Supplier No. L19549.MC

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 387824-61-5
Molecular Formula C6H5F3N2
Molecular Weight (g/mol) 162.115
MDL Number MFCD02183554
InChI Key DYAGVPKVELUYPJ-UHFFFAOYSA-N
PubChem CID 2782810
IUPAC Name 3-(trifluoromethyl)pyridin-4-amine
SMILES C1=CN=CC(=C1N)C(F)(F)F

Specifications

Melting Point 98°C to 102°C
Quantity 100 mg
UN Number UN2811
Synonym 4-amino-3-trifluoromethyl pyridine, 3-trifluoromethyl pyridin-4-amine, 4-amino-3-trifluoromethylpyridine, 4-pyridinamine, 3-trifluoromethyl, 3-trifluoromethyl-4-pyridylamine, pubchem2994, acmc-1aev4, 3-trifluoromethyl-4-pyridinamine, 4-pyridinamine,3-trifluoromethyl
Formula Weight 162.11
Percent Purity 97%
Chemical Name or Material 4-Amino-3-(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.