missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-(trifluoromethyl)benzoic acid, 98%

Product Code. 15455217
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15455217 10 g 10g
15445217 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455217 Supplier Thermo Scientific Alfa Aesar Supplier No. H32487.09
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 400-76-0 | C8H6F3NO2 | 205.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 400-76-0
Molecular Formula C8H6F3NO2
Molecular Weight (g/mol) 205.14
MDL Number MFCD03407960
InChI Key NPPPORJZPNJXNQ-UHFFFAOYSA-N
Synonym 4-amino-3-trifluoromethyl benzoic acid, 4-amino-3-trifluoromethylbenzioc acid, rarechem al bo 0877, 4-amino-5-trifluoromethylbenzoic acid, 4-amino-3-trifluoromethyl-benzoic acid, benzoic acid, 4-amino-3-trifluoromethyl, 4-amino-3-trifluoromethyl benzoic acid 3-trifluoromethyl-4-aminobenzoic acid, 4-amino-3-trifluoromethylbenzoicacid, pubchem2211
PubChem CID 3254286
IUPAC Name 4-amino-3-(trifluoromethyl)benzoic acid
SMILES NC1=C(C=C(C=C1)C(O)=O)C(F)(F)F

Specifications

Melting Point 212°C to 215°C
Quantity 10 g
Beilstein 3291493
Formula Weight 205.14
Percent Purity 98%
Chemical Name or Material 4-Amino-3-(trifluoromethyl)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.