missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-(trifluoromethoxy)benzonitrile, ≥97%, Thermo Scientific™

Product Code. 10108454
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10108454 5 g 5g
10109622 1 g 1g
10138552 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10108454 Supplier Thermo Scientific Maybridge Supplier No. TL00680DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175278-23-6
Molecular Formula C8H5F3N2O
Molecular Weight (g/mol) 202.14
MDL Number MFCD00204177
InChI Key QNJPSMLILRHZOW-UHFFFAOYSA-N
Synonym 4-amino-3-trifluoromethoxy benzonitrile, benzonitrile, 4-amino-3-trifluoromethoxy, 3-trifluoromethoxy-4-aminobenzonitrile, 4-amino-3-trifluoromethoxy benzenecarbonitrile, pubchem4688, ksc535q4r, buttpark 81\01-97, 4-cyano-2-trifluoromethoxy-aniline, 4-cyano-2-trifluoromethoxy aniline, 4-amino-3-trifluoromethoxy-benzonitrile
PubChem CID 2735952
IUPAC Name 4-amino-3-(trifluoromethoxy)benzonitrile
SMILES NC1=C(OC(F)(F)F)C=C(C=C1)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 202.13
Percent Purity 97%
Chemical Name or Material 4-Amino-3-(trifluoromethoxy)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.