missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-3-(trifluoromethoxy)benzoic acid, 95%, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 221.13 |
Percent Purity | 95% |
Chemical Name or Material | 4-Amino-3-(trifluoromethoxy)benzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'