missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-(trifluoromethoxy)benzoic acid, 95%, Thermo Scientific™

Artikelnummer. 10255704
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
Packungsgröße:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packaging unitSize
10255704 50 g Amber Glass Bottle 50g
10029511 1 g Amber Glass Bottle 1g
10410964 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10255704 Supplier Thermo Scientific Maybridge Supplier No. TL00679EE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175278-22-5
Molecular Formula C8H6F3NO3
Molecular Weight (g/mol) 221.135
InChI Key IXJFWBNYFTWBOR-UHFFFAOYSA-N
Synonym 4-amino-3-trifluoromethoxy benzoic acid, rarechem al bo 0798, buttpark 83\07-32, 4-amino-3-trifluoromethoxy-benzoic acid, 4-amine-3-trifluoromethoxy benzoic acid, benzoic acid, 4-amino-3-trifluoromethoxy, 4-amino-3-trifluoromethoxybenzoicacid, pubchem14042, acmc-209e9s
PubChem CID 2735951
IUPAC Name 4-amino-3-(trifluoromethoxy)benzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)OC(F)(F)F)N

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 50 g
Formula Weight 221.13
Percent Purity 95%
Chemical Name or Material 4-Amino-3-(trifluoromethoxy)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.