missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-3-phenylbutanoic Acid, TRC
Specifications
Specifications
| CAS | 1078-21-3 |
| Molecular Formula | C10H13NO2 |
| Synonym | -(Aminomethyl)benzenepropanoic Acid;(±)-Fenibut;(±)-beta-Phenyl-GABA;3-Phenyl-4-aminobutanoic Acid;4-Amino-3-phenylbutyric Acid;Anvifen;DL-4-Amino-3-phenylbutanoic Acid;DL-beta-Phenyl-gamma-aminobutyric Acid;Fenibut;Fenigam;Fenigama;P-GABA;PhGABA;Phenibut;Phenigam;Phenybut;Phenygam;beta-Phenyl-GABA;beta-Phenyl-gamma-aminobutyric Acid;gamma-Amino-beta-phenylbutyric Acid |
| InChI Formula | InChI=1S/C10H13NO2/c11-7-9(6-10(12)13)8-4-2-1-3-5-8/h1-5,9H,6-7,11H2,(H,12,13) |
| SMILES | NCC(CC(=O)O)c1ccccc1 |
| IUPAC Name | 4-amino-3-phenylbutanoic acid |
| Molecular Weight (g/mol) | 179.22 |
| Quantity | 1 g |
| Formula Weight | 179.0946 |
| Percent Purity | >95% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?